This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01331679_m1
See other CEP170B GE Assays ›
Gene Symbol:
Cep170b
Gene Name:
centrosomal protein 170B
Gene Aliases:
AI466840, AW555464, Kiaa0284, mKIAA0284
Chromosome Location:
Chr.12: 112720591 - 112746592 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01331679_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cep170b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001024602.3 NP_001019773.2 5 - 6 572 86
GenBank mRNA AK122236.1 - 6 - 7 501 86
AK147410.1 - 5 - 6 513 86
AK147612.1 - 3 - 4 177 86
BC050077.1 - 5 - 6 406 86
BC141174.1 - 4 - 5 372 9055786 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200001269.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cep170b

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200009131.mm.1 NM_001024602.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200009131.mm.1 NM_001024602.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200009131.mm.1 TC1200001269.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200009129.mm.1 NM_001024602.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200009129.mm.1 NM_001024602.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200009129.mm.1 TC1200001269.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200009130.mm.1 NM_001024602.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200009130.mm.1 NM_001024602.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200009130.mm.1 TC1200001269.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0