This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01331068_m1
See other ACVR1 GE Assays ›
Gene Symbol:
Acvr1
Gene Name:
activin A receptor, type 1
Gene Aliases:
ALK2, ActR-I, ActRIA, Acvr, Acvrlk2, Alk-2, Alk8, D330013D15Rik, SKR1, Tsk7L
Chromosome Location:
Chr.2: 58446438 - 58566828 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01331068_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Acvr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001110204.1 NP_001103674.1 11 - 12 1923 93
NM_001110205.1 NP_001103675.1 9 - 10 1401 93
NM_007394.3 NP_031420.2 10 - 11 1678 93
GenBank mRNA AF332087.1 - 8 - 9 1311 93
AF332088.1 - 8 - 9 1311 93
AK053428.1 - 11 - 12 1922 93
AK150014.1 - 10 - 11 1678 93
AK150075.1 - 10 - 11 1678 93
BC058718.1 - 8 - 9 1399 6418954 93
L15436.1 - 9 - 10 1751 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200003579.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Acvr1

Panther Classification:

Molecular Function -

serine/threonine protein kinase receptor

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
patterning of blood vessels
urogenital system development
in utero embryonic development
gastrulation with mouth forming second
mesoderm formation
neural crest cell migration
embryonic heart tube morphogenesis
mitral valve morphogenesis
atrial septum primum morphogenesis
protein phosphorylation
transmembrane receptor protein serine/threonine kinase signaling pathway
transforming growth factor beta receptor signaling pathway
germ cell development
determination of left/right symmetry
gastrulation
mesoderm development
heart development
embryo development
negative regulation of signal transduction
positive regulation of pathway-restricted SMAD protein phosphorylation
phosphorylation
peptidyl-threonine phosphorylation
signal transduction by protein phosphorylation
regulation of ossification
positive regulation of bone mineralization
BMP signaling pathway
activin receptor signaling pathway
negative regulation of activin receptor signaling pathway
intracellular signal transduction
positive regulation of osteoblast differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
smooth muscle cell differentiation
pharyngeal system development
pathway-restricted SMAD protein phosphorylation
cardiac muscle cell fate commitment
endocardial cushion cell fate commitment
positive regulation of determination of dorsal identity
negative regulation of extrinsic apoptotic signaling pathway
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
transmembrane receptor protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
transforming growth factor beta receptor activity, type I
ATP binding
kinase activity
activin receptor activity, type I
transferase activity
transferase activity, transferring phosphorus-containing groups
growth factor binding
protein homodimerization activity
SMAD binding
metal ion binding
activin binding
transforming growth factor beta binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200030475.mm.1 NM_001110204.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200030475.mm.1 NM_001110204.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200030475.mm.1 NM_001110205.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200030475.mm.1 NM_001110205.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200030475.mm.1 NM_007394.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200030475.mm.1 NM_007394.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200030475.mm.1 TC0200003579.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0