This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01330918_g1
See other PPP6R1 GE Assays ›
Gene Symbol:
Ppp6r1
Gene Name:
protein phosphatase 6, regulatory subunit 1
Gene Aliases:
2010309P17Rik, AI836219, B430201G11Rik, D030074N20Rik, Pp6r1, Saps1, mKIAA1115
Chromosome Location:
Chr.7: 4631495 - 4658950 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01330918_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp6r1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172894.2 NP_766482.2 11 - 12 1837 75
GenBank mRNA AK122450.1 - 7 - 8 2940 75
BC053076.1 - 11 - 12 1837 5698212 75
BC141039.1 - 10 - 11 1384 9055651 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002166.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp6r1

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

regulation of phosphoprotein phosphatase activity
protein phosphatase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700019768.mm.1 NM_172894.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700019768.mm.1 NM_172894.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700019768.mm.1 TC0700002166.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700019769.mm.1 NM_172894.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700019769.mm.1 NM_172894.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700019769.mm.1 TC0700002166.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0