This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01330325_g1
See other GNPTG GE Assays ›
Gene Symbol:
Gnptg
Gene Name:
N-acetylglucosamine-1-phosphotransferase, gamma subunit
Gene Aliases:
6430527N14Rik, A830081F19, AU067667, AU067744, Mdcp1, Tce7
Chromosome Location:
Chr.17: 25234317 - 25240170 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01330325_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gnptg
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172529.3 NP_766117.2 6 - 7 413 91
GenBank mRNA AK044021.1 - 5 - 6 493 91
AY465529.1 - 6 - 7 421 91
BC023869.1 - 4 - 5 563 91
BC055872.1 - 6 - 7 413 4925332 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700001655.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gnptg

Panther Classification:

Molecular Function -

protein-binding activity modulator

Gene Ontology Categories:

Function(s) Process(es)

N-glycan processing to lysosome
carbohydrate phosphorylation
UDP-N-acetylglucosamine-lysosomal-enzyme N-acetylglucosaminephosphotransferase activity
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700014984.mm.1 NM_172529.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700014984.mm.1 NM_172529.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700014984.mm.1 TC1700001655.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0