This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01329578_g1
See other SLC4A11 GE Assays ›
Gene Symbol:
Slc4a11
Gene Name:
solute carrier family 4, sodium bicarbonate transporter-like, member 11
Gene Aliases:
AI503023, BTR1, NaBC1
Chromosome Location:
Chr.2: 130684107 - 130697519 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01329578_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc4a11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081162.1 NP_001074631.1 10 - 11 1287 60
GenBank mRNA AK052665.1 - 5 - 6 2156 60
AK158115.1 - 10 - 11 1241 60
BC036154.1 - 1 - 2 786 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
sodium ion transport
anion transport
bicarbonate transport
proton transport
cellular cation homeostasis
borate transmembrane transport
sodium ion transmembrane transport
fluid transport
borate transport
ion homeostasis
regulation of intracellular pH
transporter activity
sodium channel activity
inorganic anion exchanger activity
anion transmembrane transporter activity
bicarbonate transmembrane transporter activity
hydrogen ion channel activity
symporter activity
anion:anion antiporter activity
borate transmembrane transporter activity
protein dimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0