This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01329181_m1
See other DPP4 GE Assays ›
Gene Symbol:
Dpp4
Gene Name:
dipeptidylpeptidase 4
Gene Aliases:
Cd26, Dpp-4, THAM
Chromosome Location:
Chr.2: 62330073 - 62412317 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01329181_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dpp4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159543.1 NP_001153015.1 11 - 12 1447 89
NM_010074.3 NP_034204.1 11 - 12 1447 89
GenBank mRNA AK085370.1 - 11 - 12 1030 89
AK153939.1 - 11 - 12 1061 89
AK163085.1 - 11 - 12 1452 89
AK164775.1 - 11 - 12 1030 89
X58384.1 - 11 - 12 1124 89

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dpp4

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

behavioral fear response
response to hypoxia
regulation of T cell mediated immunity
proteolysis
cell adhesion
positive regulation of cell proliferation
negative regulation of extracellular matrix disassembly
T cell costimulation
regulation of cell-cell adhesion mediated by integrin
locomotory exploration behavior
psychomotor behavior
T cell activation
endothelial cell migration
viral entry into host cell
establishment of localization
virus receptor activity
protease binding
aminopeptidase activity
serine-type endopeptidase activity
receptor binding
collagen binding
peptidase activity
serine-type peptidase activity
dipeptidyl-peptidase activity
hydrolase activity
peptide binding
identical protein binding
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0