This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01329177_m1
See other FAP GE Assays ›
Gene Symbol:
Fap
Gene Name:
fibroblast activation protein
Gene Aliases:
SIMP
Chromosome Location:
Chr.2: 62500936 - 62575508 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01329177_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fap
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007986.3 NP_032012.1 6 - 7 581 76
GenBank mRNA AK051959.1 - 6 - 7 484 76
AK136578.1 - 5 - 6 571 76
BC019190.1 - 6 - 7 485 5028579 76
Y10007.1 - 6 - 7 581 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200003629.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fap

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
proteolysis
apoptotic process
cell adhesion
regulation of collagen catabolic process
negative regulation of extracellular matrix disassembly
endothelial cell migration
proteolysis involved in cellular protein catabolic process
negative regulation of cell proliferation involved in contact inhibition
positive regulation of cell cycle arrest
mitotic cell cycle arrest
melanocyte proliferation
positive regulation of execution phase of apoptosis
melanocyte apoptotic process
negative regulation of extracellular matrix organization
protease binding
endopeptidase activity
serine-type endopeptidase activity
integrin binding
peptidase activity
serine-type peptidase activity
dipeptidyl-peptidase activity
hydrolase activity
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0