This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01328882_mH
See other PPIG GE Assays ›
Gene Symbol:
Ppig
Gene Name:
peptidyl-prolyl isomerase G (cyclophilin G)
Gene Aliases:
AU019516, AU022200, B230312B02Rik, CYP, SRCyp
Chromosome Location:
Chr.2: 69722617 - 69754059 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01328882_mH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppig
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081086.1 NP_001074555.1 9 - 10 904 93
GenBank mRNA AK134121.1 - 9 - 10 906 93
AK135178.1 - 10 - 11 1036 93
BC016126.1 - 9 - 10 766 93
BC027420.1 - 9 - 10 898 93
BC038818.1 - 9 - 10 720 93
BC094926.1 - 9 - 10 758 93
BC150694.1 - 10 - 11 925 9087605 93

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein peptidyl-prolyl isomerization
protein folding
peptidyl-prolyl cis-trans isomerase activity
isomerase activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200007944.mm.1 NM_001081086.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200007944.mm.1 NM_001081086.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200007944.mm.1 TC0200001009.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200007945.mm.1 NM_001081086.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200007945.mm.1 NM_001081086.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200007945.mm.1 TC0200001009.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200007948.mm.1 NM_001081086.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200007948.mm.1 NM_001081086.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200007948.mm.1 TC0200001009.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200007950.mm.1 NM_001081086.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200007950.mm.1 NM_001081086.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200007950.mm.1 TC0200001009.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200007949.mm.1 NM_001081086.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200007949.mm.1 NM_001081086.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200007949.mm.1 TC0200001009.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200007951.mm.1 NM_001081086.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200007951.mm.1 NM_001081086.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200007951.mm.1 TC0200001009.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200007952.mm.1 NM_001081086.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200007952.mm.1 NM_001081086.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200007952.mm.1 TC0200001009.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0