This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01328876_g1
See other PPIG GE Assays ›
Gene Symbol:
Ppig
Gene Name:
peptidyl-prolyl isomerase G (cyclophilin G)
Gene Aliases:
AU019516, AU022200, B230312B02Rik, CYP, SRCyp
Chromosome Location:
Chr.2: 69722617 - 69754059 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01328876_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppig
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081086.1 NP_001074555.1 2 - 3 348 90
GenBank mRNA AK088007.1 - 2 - 3 159 89
AK134121.1 - 2 - 3 349 90
AK135178.1 - 3 - 4 480 90
BC016126.1 - 2 - 3 210 89
BC027420.1 - 2 - 3 342 89
BC038818.1 - 2 - 3 166 89
BC048905.1 - 2 - 3 209 90
BC094926.1 - 2 - 3 202 89
BC150694.1 - 3 - 4 369 9087605 89
U91923.1 - 2 - 3 154 89

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK134121.1, BC016126.1, BC027420.1, BC048905.1, BC094926.1, BC150694.1, NM_001081086.1, U91923.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein peptidyl-prolyl isomerization
protein folding
peptidyl-prolyl cis-trans isomerase activity
isomerase activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200007930.mm.1 NM_001081086.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200007930.mm.1 NM_001081086.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200007930.mm.1 TC0200001009.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200007931.mm.1 NM_001081086.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200007931.mm.1 NM_001081086.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200007931.mm.1 TC0200001009.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0