This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01328526_m1
See other CSRP2BP GE Assays ›
Gene Symbol:
Csrp2bp
Gene Name:
cysteine and glycine-rich protein 2 binding protein
Gene Aliases:
2510008M08Rik, ATAC2, AU023459, D2Ertd473e, D2Wsu131e, E430020F17
Chromosome Location:
Chr.2: 144369032 - 144407675 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01328526_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Csrp2bp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_181417.3 NP_852082.2 2 - 3 785 124
GenBank mRNA BC031563.1 - 2 - 3 785 4949236 124

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002146.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Csrp2bp

Gene Ontology Categories:

Function(s) Process(es)

G2/M transition of mitotic cell cycle
embryo development
histone acetylation
histone H3 acetylation
histone acetyltransferase activity
protein binding
N-acetyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200016894.mm.1 NM_001166640.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200016894.mm.1 NM_001166640.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200016894.mm.1 NM_181417.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200016894.mm.1 NM_181417.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200016894.mm.1 TC0200002146.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200016891.mm.1 NM_181417.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200016891.mm.1 NM_181417.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200016891.mm.1 TC0200002146.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0