This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01327663_m1
See other KIF18A GE Assays ›
Gene Symbol:
Kif18a
Gene Name:
kinesin family member 18A
Gene Aliases:
AU024633, B130001M12Rik
Chromosome Location:
Chr.2: 109280738 - 109341749 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01327663_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Kif18a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_139303.1 NP_647464.1 9 - 10 1454 105
GenBank mRNA AK036722.1 - 9 - 10 1341 105
AK041469.1 - 9 - 10 1353 105
AK044795.1 - 9 - 10 1349 105
AK049197.1 - 9 - 10 1404 105
AK164649.1 - 9 - 10 1349 105
BC004740.1 - 8 - 9 1309 105
BC016095.1 - 9 - 10 1454 4506052 105

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200001620.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Kif18a

Panther Classification:

Molecular Function -

microtubule binding motor protein

Gene Ontology Categories:

Function(s) Process(es)

mitotic sister chromatid segregation
transport
microtubule-based movement
microtubule depolymerization
mitotic metaphase plate congression
metabolic process
protein transport
nucleotide binding
microtubule motor activity
actin binding
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, plus-end-directed
ubiquitin binding
microtubule plus-end binding
tubulin-dependent ATPase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200012089.mm.1 NM_139303.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200012089.mm.1 NM_139303.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200012089.mm.1 TC0200001620.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200012090.mm.1 NM_139303.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200012090.mm.1 NM_139303.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200012090.mm.1 TC0200001620.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0