This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01327440_m1
See other MAPKAP1 GE Assays ›
Gene Symbol:
Mapkap1
Gene Name:
mitogen-activated protein kinase associated protein 1
Gene Aliases:
AI591529, D230039K05Rik, Sin1
Chromosome Location:
Chr.2: 34406771 - 34624958 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01327440_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mapkap1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290625.1 NP_001277554.1 2 - 3 406 71
NM_177345.4 NP_796319.1 2 - 3 732 71
GenBank mRNA AK052045.1 - 2 - 3 400 71
AK132263.1 - 2 - 3 396 71
AK148428.1 - 2 - 3 721 71
BC043296.1 - 2 - 3 631 71
BC090644.1 - 1 - 2 306 30629263 71
BC096618.1 - 2 - 3 705 6392317 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000612.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mapkap1

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of Ras protein signal transduction
protein binding
phosphatidylinositol-4,5-bisphosphate binding
phosphatidylinositol-3,4,5-trisphosphate binding
Ras GTPase binding
protein kinase binding
phosphatidylinositol-3,4-bisphosphate binding
phosphatidic acid binding
phosphatidylinositol-3,5-bisphosphate binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200005631.mm.1 NM_177345.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200005631.mm.1 TC0200000612.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header