This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01327438_g1
See other SH3GLB2 GE Assays ›
Gene Symbol:
Sh3glb2
Gene Name:
SH3-domain GRB2-like endophilin B2
Gene Aliases:
SPAS-1
Chromosome Location:
Chr.2: 30344777 - 30359316 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01327438_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sh3glb2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289709.1 NP_001276638.1 7 - 8 759 53
NM_139302.2 NP_647463.1 7 - 8 759 53
GenBank mRNA AF426314.1 - 7 - 8 656 53
AK152409.1 - 7 - 8 689 53
AK155248.1 - 7 - 8 684 53
AK167762.1 - 7 - 8 732 53
BC024477.1 - 7 - 8 674 4977001 53
EF676083.1 - 7 - 8 735 53

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200003269.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sh3glb2

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200027632.mm.1 NM_001289709.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200027632.mm.1 NM_001289710.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200027632.mm.1 NM_139302.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200027632.mm.1 TC0200003269.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0