Assay Details
| Gene Symbol: F12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AW111733.1 | - | 2 - 3 | 86 | ||
| BC037085.1 | - | 12 - 13 | 1487 | 86 | ||
| BC049867.1 | - | 12 - 13 | 1510 | 86 | ||
| BC057921.1 | - | 12 - 13 | 1498 | 5101105 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

F12
Genomic Map