This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01326348_m1
See other ETL4 GE Assays ›
Gene Symbol:
Etl4
Gene Name:
enhancer trap locus 4
Gene Aliases:
6620402G01Rik, 9430077C05Rik, BC026657, E330027G05Rik, Etl-4, Skt
Chromosome Location:
Chr.2: 19909763 - 20810535 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01326348_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Etl4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001177630.2 NP_001171101.2 1 - 2 415 103
NM_001177631.1 NP_001171102.1 1 - 2 415 103
NM_029895.4 NP_084171.2 2 - 3 628 103
NM_178059.5 NP_835160.2 2 - 3 628 103
GenBank mRNA AB093289.2 - 1 - 2 205 103
AB125594.2 - 2 - 3 628 103
AB125595.2 - 2 - 3 628 103
AK052733.1 - 1 - 2 103 103
AK147510.1 - 1 - 2 322 103
BC050016.1 - 1 - 2 118 103
BC158051.1 - 1 - 2 137 9007405 103
BC172143.1 - 1 - 2 137 103

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000319.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Etl4

Gene Ontology Categories:

Function(s)

multicellular organism development
embryonic skeletal system development

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 831221 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0