This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01325745_m1
See other GUCY2F GE Assays ›
Gene Symbol:
Gucy2f
Gene Name:
guanylate cyclase 2f
Gene Aliases:
A930002I06, GC-F, GC2
Chromosome Location:
Chr.X: 142079286 - 142197276 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01325745_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gucy2f
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001007576.2 NP_001007577.1 11 - 12 2520 86
GenBank mRNA AK044234.1 - 11 - 12 2521 86
AY651761.1 - 11 - 12 2515 86
BC115714.1 - 10 - 11 2350 40085764 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00003096.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gucy2f

Panther Classification:

Molecular Function -

guanylate cyclase

Gene Ontology Categories:

Function(s) Process(es)

cGMP biosynthetic process
protein phosphorylation
visual perception
cyclic nucleotide biosynthetic process
intracellular signal transduction
response to stimulus
nucleotide binding
guanylate cyclase activity
protein kinase activity
ATP binding
GTP binding
transferase activity, transferring phosphorus-containing groups
lyase activity
phosphorus-oxygen lyase activity
protein homodimerization activity
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00019450.mm.1 NM_001007576.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00019450.mm.1 NM_001007576.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00019450.mm.1 TC0X00003096.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00019451.mm.1 NM_001007576.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00019451.mm.1 NM_001007576.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00019451.mm.1 TC0X00003096.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0