This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01325453_m1
See other DZANK1 GE Assays ›
Gene Symbol:
Dzank1
Gene Name:
double zinc ribbon and ankyrin repeat domains 1
Gene Aliases:
2810039F03Rik, 6330439K17Rik, 6430408C01, AI851115, Ankrd64
Chromosome Location:
Chr.2: 144470555 - 144527425 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01325453_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dzank1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172859.2 NP_766447.2 1 - 2 115 109
GenBank mRNA AK012889.1 - 1 - 2 82 109
AK032182.1 - 1 - 2 117 109
AK032632.1 - 1 - 2 106 109
AK163994.1 - 1 - 2 106 109
BC089524.1 - 1 - 2 82 109

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK032182.1, AK032632.1, BC089524.1, NM_172859.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dzank1

Gene Ontology Categories:

Function(s) Process(es)

mitophagy in response to mitochondrial depolarization
molecular_function
zinc ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200040801.mm.1 NM_172859.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200040801.mm.1 NM_172859.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200040801.mm.1 TC0200004762.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200040804.mm.1 NM_172859.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200040804.mm.1 NM_172859.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200040804.mm.1 TC0200004762.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200040805.mm.1 NM_172859.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200040805.mm.1 NM_172859.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200040805.mm.1 TC0200004762.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0