This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01325389_m1
See other WWC1 GE Assays ›
Gene Symbol:
Wwc1
Gene Name:
WW, C2 and coiled-coil domain containing 1
Gene Aliases:
AA408228, AU017197, BC037006, Kibra, mKIAA0869
Chromosome Location:
Chr.11: 35839178 - 35980089 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01325389_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Wwc1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_170779.1 NP_740749.1 15 - 16 2283 59
GenBank mRNA AB271907.1 - 15 - 16 2253 59
AK220259.1 - 2654 59
BC006733.1 - 12 - 13 1872 59
BC017638.1 - 8 - 9 1409 59
BC037006.1 - 7 - 8 1131 5116674 59
DQ256090.1 - 15 - 16 2283 59

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Wwc1

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
cell migration
regulation of hippo signaling
negative regulation of hippo signaling
positive regulation of MAPK cascade
negative regulation of organ growth
transcription coactivator activity
kinase binding
protein binding, bridging
protein complex scaffold

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0