This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01324436_m1
See other POMT1 GE Assays ›
Gene Symbol:
Pomt1
Gene Name:
protein-O-mannosyltransferase 1
Gene Aliases:
AI505244
Chromosome Location:
Chr.2: 32236595 - 32255005 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01324436_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pomt1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145145.1 NP_660127.1 17 - 18 1793 69
GenBank mRNA AK053889.1 - 16 - 17 2313 69
AK134770.1 - 17 - 18 1837 69
AK153984.1 - 17 - 18 1892 69
AY007238.1 - 17 - 18 1833 69
AY494857.1 - 17 - 18 1853 69
BC027325.1 - 17 - 18 1793 3981366 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000572.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pomt1

Panther Classification:

Molecular Function -

transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein O-linked glycosylation
extracellular matrix organization
mannosyltransferase activity
dolichyl-phosphate-mannose-protein mannosyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200005087.mm.1 NM_145145.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005087.mm.1 NM_145145.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005087.mm.1 TC0200000572.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0