This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01324332_m1
See other HDGFRP3 GE Assays ›
Gene Symbol:
Hdgfrp3
Gene Name:
hepatoma-derived growth factor, related protein 3
Gene Aliases:
2700022B06Rik, HRP-3
Chromosome Location:
Chr.7: 81881256 - 81934459 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01324332_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hdgfrp3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013886.4 NP_038914.2 4 - 5 894 126
GenBank mRNA AB029493.1 - 4 - 5 457 126
AK014849.1 - 4 - 5 718 126
AK019487.1 - 4 - 5 557 126
AK044858.1 - 4 - 5 719 126
AK051401.1 - 4 - 5 816 126
AK162418.1 - 4 - 5 537 126
BC055734.1 - 4 - 5 481 6810355 126

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700003622.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hdgfrp3

Panther Classification:

Molecular Function -

transcription cofactor

Gene Ontology Categories:

Function(s) Process(es)

cell proliferation
chromatin binding
growth factor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700030841.mm.1 NM_013886.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700030841.mm.1 NM_013886.4 Refseq
Affymetrix Mouse Clariom S Array PSR0700030841.mm.1 TC0700003622.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0