This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01323795_m1
See other ACIN1 GE Assays ›
Gene Symbol:
Acin1
Gene Name:
apoptotic chromatin condensation inducer 1
Gene Aliases:
2610036I19Rik, 2610510L13Rik, Acinus, Acn, C79325, acinusL, acinusS, mKIAA0670
Chromosome Location:
Chr.14: 54642161 - 54686931 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01323795_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Acin1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001085473.2 NP_001078942.2 5 - 6 849 78
NM_023190.3 NP_075679.2 5 - 6 849 78
GenBank mRNA AK011698.1 - 5 - 6 811 78
AK030449.1 - 5 - 6 542 78
AK050467.1 - 4 - 5 412 78
BC075691.1 - 5 - 6 534 78

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Acin1

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of defense response to virus by host
mRNA processing
apoptotic process
RNA splicing
regulation of autophagy
apoptotic chromosome condensation
regulation of apoptotic process
positive regulation of apoptotic process
negative regulation of mRNA splicing, via spliceosome
mitophagy in response to mitochondrial depolarization
nucleotide binding
nucleic acid binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0