This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01322817_m1
See other CDC42BPA GE Assays ›
Gene Symbol:
Cdc42bpa
Gene Name:
CDC42 binding protein kinase alpha
Gene Aliases:
A930014J19Rik, DMPK-like
Chromosome Location:
Chr.1: 179960093 - 180165606 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01322817_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdc42bpa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033285.1 NP_001028457.1 6 - 7 709 98
GenBank mRNA AK138649.1 - 7 - 8 1198 98
BC158017.1 - 6 - 7 709 9007323 98
BC158095.1 - 6 - 7 709 9007382 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100001703.mm.1

Set Membership:

Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

microtubule cytoskeleton organization
protein phosphorylation
cytoskeleton organization
nuclear migration
phosphorylation
cell migration
actomyosin structure organization
actin cytoskeleton reorganization
intracellular signal transduction
regulation of small GTPase mediated signal transduction
nucleotide binding
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
identical protein binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100014091.mm.1 NM_001033285.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100014091.mm.1 NM_001033285.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100014091.mm.1 TC0100001703.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0