This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01321631_m1
See other CAPN2 GE Assays ›
Gene Symbol:
Capn2
Gene Name:
calpain 2
Gene Aliases:
AI326419, Capa-2, Capa2, m-calpain, m-calpin
Chromosome Location:
Chr.1: 182467259 - 182517483 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01321631_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Capn2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009794.3 NP_033924.2 15 - 16 1824 69
GenBank mRNA AF015038.1 - 15 - 16 1770 69
AK052141.1 - 13 - 14 1412 69
AK150695.1 - 15 - 16 1798 69
AK152094.1 - 15 - 16 1798 69
AK152790.1 - 15 - 16 1801 69
AK152994.1 - 15 - 16 1801 69
AK154855.1 - 15 - 16 1838 69
AK199295.1 - 4 - 5 260 69
BC054726.1 - 15 - 16 1824 5703563 69
D38117.1 - 15 - 16 1865 69
Y10139.1 - 15 - 16 1706 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100003715.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Capn2

Panther Classification:

Molecular Function -

cysteine protease

Gene Ontology Categories:

Function(s) Process(es)

blastocyst development
proteolysis
myoblast fusion
protein autoprocessing
regulation of cytoskeleton organization
cellular response to amino acid stimulus
calcium-dependent cysteine-type endopeptidase activity
calcium ion binding
cytoskeletal protein binding
peptidase activity
cysteine-type peptidase activity
hydrolase activity
metal ion binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100030441.mm.1 NM_009794.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100030441.mm.1 NM_009794.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100030441.mm.1 TC0100003715.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0