This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01321306_m1
See other LOXL2 GE Assays ›
Gene Symbol:
Loxl2
Gene Name:
lysyl oxidase-like 2
Gene Aliases:
1110004B06Rik, 4930526G11Rik, 9430067E15Rik
Chromosome Location:
Chr.14: 69608692 - 69695834 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01321306_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Loxl2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK034957.1 - 1 - 2 125 74
AK145323.1 - 1 - 2 125 74
AK159386.1 - 1 - 2 121 74
BC086801.1 - 223 30605240 74

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK034957.1, AK145323.1, AK159386.1, BC086801.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

oxidase

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400007817.mm.1 TC1400000976.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400007816.mm.1 TC1400000976.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400007820.mm.1 TC1400000976.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400007818.mm.1 TC1400000976.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400007819.mm.1 TC1400000976.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400007821.mm.1 TC1400000976.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 374833 - -
ABI Mouse Genome Survey Microarray 418310 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header