This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01321282_g1
See other CCAR2 GE Assays ›
Gene Symbol:
Ccar2
Gene Name:
cell cycle activator and apoptosis regulator 2
Gene Aliases:
2610301G19Rik, mKIAA1967
Chromosome Location:
Chr.14: 70138168 - 70153853 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01321282_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ccar2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_146055.3 NP_666167.3 5 - 6 557 62
GenBank mRNA AK028194.1 - 5 - 6 479 62
AK031472.1 - 1 - 2 97 62
AK050238.1 - 5 - 6 557 62
BC021475.1 - 5 - 6 587 5343456 62
BC038346.1 - 5 - 6 509 3985250 62

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK031472.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ccar2

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
mRNA processing
apoptotic process
cellular response to DNA damage stimulus
cell cycle
RNA splicing
response to UV
regulation of protein stability
negative regulation of proteasomal ubiquitin-dependent protein catabolic process
regulation of DNA-templated transcription, elongation
regulation of circadian rhythm
positive regulation of apoptotic process
negative regulation of catalytic activity
mitochondrial fragmentation involved in apoptotic process
rhythmic process
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
positive regulation of DNA damage checkpoint
RNA polymerase II core binding
enzyme inhibitor activity
protein binding
enzyme binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400018082.mm.1 NM_146055.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400018082.mm.1 NM_146055.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400018082.mm.1 TC1400002331.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0