Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400018088.mm.1 |
NM_146055.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018088.mm.1 |
NM_146055.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018088.mm.1 |
TC1400002331.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400018085.mm.1 |
NM_146055.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018085.mm.1 |
NM_146055.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018085.mm.1 |
TC1400002331.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400018086.mm.1 |
NM_146055.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018086.mm.1 |
NM_146055.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018086.mm.1 |
TC1400002331.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400018087.mm.1 |
NM_146055.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018087.mm.1 |
NM_146055.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018087.mm.1 |
TC1400002331.mm.1 |
MTA TC |