This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01320557_g1
See other SEMA4B GE Assays ›
Gene Symbol:
Sema4b
Gene Name:
sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4B
Gene Aliases:
SemC, Semac, mKIAA1745
Chromosome Location:
Chr.7: 80186841 - 80226527 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01320557_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sema4b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013659.4 NP_038687.2 11 - 12 1645 73
GenBank mRNA AK028896.1 - 11 - 12 1608 73
AK030331.1 - 10 - 11 1645 73
AK134973.1 - 10 - 11 1576 73
AK145570.1 - 11 - 12 1646 73
AK154969.1 - 10 - 11 1426 73
AK173249.1 - 11 - 12 1593 73
DQ073331.1 - 10 - 11 1401 73
X85992.1 - 9 - 10 1237 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700001145.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sema4b

Panther Classification:

Molecular Function -

membrane-bound signaling molecule intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
nervous system development
cell differentiation
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700010041.mm.1 NM_013659.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700010041.mm.1 NM_013659.4 Refseq
Affymetrix Mouse Clariom S Array PSR0700010041.mm.1 TC0700001145.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0