This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01320171_m1
See other 1700109G14RIK GE Assays ›
Gene Symbol:
1700109G14Rik
Gene Name:
RIKEN cDNA 1700109G14 gene
Gene Aliases:
1700126G13Rik
Chromosome Location:
Chr.14: 61292983 - 61305334 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01320171_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1700109G14Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_033788.1 1 - 2 195 95
GenBank mRNA AK007149.1 - 1 - 2 195 95
AK018935.1 - 1 - 2 158 95
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY707233.1 - 1 - 2 195 Chr. 14 - 61294630 - 61305334 [-] on Build GRCm38 95
GenBankmRNA CN834495.1 - 172 Chr. - - [-] on Build 95
GenBankmRNA BU935660.1 - 1 - 2 151 Chr. 14 - 61292980 - 61305289 [-] on Build GRCm38 95
GenBankmRNA CF106243.1 - 1 - 2 121 Chr. 14 - 61293140 - 61305260 [-] on Build GRCm38 95
GenBankmRNA CN838553.1 - 126 Chr. - - [-] on Build 95
GenBankmRNA BY707180.1 - 1 - 2 158 Chr. 14 - 61293087 - 61305296 [-] on Build GRCm38 95

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream sarcoglycan,
gamma (dystrophin-associated glycoprotein)
Sgcg 35kDa,
5430420E18Rik,
AI642964,
gamma-SG
-19.8kb Sense Chr. 14 - 61219106 - 61274858 on NCBI Build GRCm38
Downstream ADP-ribosylation factor-like 11 Arl11 ARLTS1,
C730007L20Rik
992.0 Anti-sense Chr. 14 - 61306288 - 61311937 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400002184.mm.1

Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400016920.mm.1 NR_033788.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400016920.mm.1 NR_033788.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400016920.mm.1 TC1400002184.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400016923.mm.1 NR_033788.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400016923.mm.1 NR_033788.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400016923.mm.1 TC1400002184.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400016922.mm.1 NR_033788.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400016922.mm.1 NR_033788.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400016922.mm.1 TC1400002184.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0