This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01319791_m1
See other D030025E07RIK GE Assays ›
Gene Symbol:
D030025E07Rik
Gene Name:
RIKEN cDNA D030025E07 gene
Gene Aliases:
-
Chromosome Location:
Chr.3: 128117015 - 128231605 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01319791_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: D030025E07Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045704.1 2 - 3 382 148
GenBank mRNA AK050844.1 - 2 - 3 383 148
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB655047.1 - 2 - 3 383 Chr. 3 - 128124586 - 128231603 [-] on Build GRCm38 148

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream RIKEN cDNA 9830132P13 gene 9830132P13Rik 9830132P13 -194.6kb Anti-sense Chr. 3 - 127916171 - 127955220 on NCBI Build GRCm38
Downstream paired-like homeodomain transcription factor 2 Pitx2 9430085M16Rik,
Brx1,
Brx1b,
Munc30,
Otlx2,
Ptx2,
Rieg
988.3kb Anti-sense Chr. 3 - 129199878 - 129219594 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002921.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300023100.mm.1 NR_045704.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300023100.mm.1 NR_045704.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300023100.mm.1 TC0300002921.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300023103.mm.1 NR_045704.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300023103.mm.1 NR_045704.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300023103.mm.1 TC0300002921.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300023104.mm.1 NR_045704.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300023104.mm.1 NR_045704.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300023104.mm.1 TC0300002921.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300023106.mm.1 NR_045704.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300023106.mm.1 NR_045704.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300023106.mm.1 TC0300002921.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300023102.mm.1 NR_045704.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300023102.mm.1 NR_045704.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300023102.mm.1 TC0300002921.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300023105.mm.1 NR_045704.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300023105.mm.1 NR_045704.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300023105.mm.1 TC0300002921.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0