This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01319229_g1
See other RIPK3 GE Assays ›
Gene Symbol:
Ripk3
Gene Name:
receptor-interacting serine-threonine kinase 3
Gene Aliases:
2610528K09Rik, AW107945, Rip3
Chromosome Location:
Chr.14: 55784995 - 55788857 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01319229_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ripk3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001164107.1 NP_001157579.1 3 - 4 675 68
NM_001164108.1 NP_001157580.1 3 - 4 773 68
NM_019955.2 NP_064339.2 3 - 4 692 68
GenBank mRNA AF178953.1 - 3 - 4 621 68
AK012177.1 - 3 - 4 675 68
AK154505.1 - 3 - 4 685 68
BC029210.1 - 3 - 4 643 3590770 68

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ripk3

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

regulation of T cell mediated cytotoxicity
regulation of adaptive immune response
protein phosphorylation
I-kappaB kinase/NF-kappaB signaling
positive regulation of phosphatase activity
programmed cell death
viral process
phosphorylation
signal transduction by protein phosphorylation
activation of protein kinase activity
regulation of interferon-gamma production
T cell differentiation in thymus
NIK/NF-kappaB signaling
T cell homeostasis
regulation of activated T cell proliferation
protein autophosphorylation
lymph node development
spleen development
thymus development
positive regulation of NF-kappaB transcription factor activity
protein homooligomerization
protein heterooligomerization
positive regulation of transferase activity
positive regulation of ligase activity
positive regulation of oxidoreductase activity
positive regulation of necroptotic process
regulation of activation-induced cell death of T cells
necroptotic process
positive regulation of protein deacetylation
amyloid fibril formation
regulation of reactive oxygen species metabolic process
positive regulation of reactive oxygen species metabolic process
regulation of CD8-positive, alpha-beta cytotoxic T cell extravasation
positive regulation of intrinsic apoptotic signaling pathway
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
NF-kappaB-inducing kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein complex binding
identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0