This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01319067_m1
See other PPP2R2A GE Assays ›
Gene Symbol:
Ppp2r2a
Gene Name:
protein phosphatase 2, regulatory subunit B, alpha
Gene Aliases:
2410004D02Rik, mKIAA1541
Chromosome Location:
Chr.14: 67014056 - 67072471 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01319067_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp2r2a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001205188.1 NP_001192117.1 9 - 10 1439 129
GenBank mRNA AK010380.1 - 9 - 10 1404 129

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400002283.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Protein Phosphatase 3' Most Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
signal transduction
response to morphine
protein serine/threonine phosphatase activity
protein phosphatase type 2A regulator activity
protein complex binding
tau protein binding
protein phosphatase 2A binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400017630.mm.1 NM_001205188.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017630.mm.1 NM_001205188.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017630.mm.1 NM_028032.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017630.mm.1 NM_028032.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400017630.mm.1 TC1400002283.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 303226 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0