This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01318653_m1
See other BC046251 GE Assays ›
Gene Symbol:
BC046251
Gene Name:
cDNA sequence BC046251
Gene Aliases:
-
Chromosome Location:
Chr.7: 65582031 - 65619342 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01318653_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BC046251
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK149279.1 - 5 - 6 2554 73
BC046251.1 - 7 - 8 1146 73

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream tight junction protein 1 Tjp1 ZO1 -83.4kb Anti-sense Chr. 7 - 65296165 - 65527806 on NCBI Build GRCm38
Downstream threonyl-tRNA synthetase-like 2 Tarsl2 A530046H20Rik 26.0kb Sense Chr. 7 - 65644898 - 65692093 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 192156 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 120735 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 167438 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 575758 AF047699.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 575758 AF055573.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 575758 BC012662.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 575758 NM_010210.2 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 925328 AB052157.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 925328 AF345951.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 925328 AF345952.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 925328 AF345953.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 925328 AF345954.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 925328 AK014441.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 925328 BC057321.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 925328 BC059232.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 925328 NM_001048054.1 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 925328 NM_130447.2 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 783788 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 776019 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 516050 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 189149 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 891070 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 558910 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 23155 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 751109 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 863628 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 3629 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 75904 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 78908 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06221934 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06221882 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01533659 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119044 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06221882 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183604 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183505 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183474 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183571 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183534 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06109759 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183562 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183513 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183573 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183500 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183495 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183560 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183598 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183607 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183504 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183563 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06112301 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183490 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183559 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183516 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183473 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183508 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06348079 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183605 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183520 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06587000 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183514 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183493 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183527 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183476 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183465 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183484 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183566 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183596 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06109760 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183558 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183491 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06091072 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183506 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06240243 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183522 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183523 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183488 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183467 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183569 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183529 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183478 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183510 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183565 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183572 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06116184 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183511 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183472 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183481 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183475 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183531 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06036283 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06389176 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183480 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183530 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183602 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183525 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183507 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183518 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183502 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06081438 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05918535 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183561 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183521 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183499 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183606 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06068983 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25183485 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06472698 - -
Array Name Probe IDs Targets Target Type
Mouse Genome 430 2.0 Array 1459653_PM_at - -
Mouse Genome 430 2.0 Array 1459653_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0