This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01318594_g1
See other TICRR GE Assays ›
Gene Symbol:
Ticrr
Gene Name:
TOPBP1-interacting checkpoint and replication regulator
Gene Aliases:
5730590G19Rik, BC028450, Lrrk1
Chromosome Location:
Chr.7: 79660196 - 79698148 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01318594_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ticrr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029835.1 NP_084111.1 11 - 12 2477 86
GenBank mRNA AK041726.1 - 11 - 12 2427 86
AK051639.1 - 11 - 12 2440 86
AK054492.1 - 11 - 12 2427 86
AK137597.1 - 11 - 12 2482 86
AK157802.1 - 10 - 11 4034 86
BC158103.1 - 11 - 12 2441 9088180 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700001131.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ticrr

Gene Ontology Categories:

Function(s) Process(es)

DNA replication
DNA repair
cellular response to DNA damage stimulus
cell cycle
response to ionizing radiation
regulation of DNA-dependent DNA replication initiation
mitotic DNA replication checkpoint
chromatin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700009949.mm.1 NM_029835.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700009949.mm.1 NM_029835.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700009949.mm.1 TC0700001131.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0