Assay Details
Gene Symbol: Ticrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029835.1 | NP_084111.1 | 9 - 10 | 2304 | 69 | |
GenBank mRNA | AK041726.1 | - | 9 - 10 | 2254 | 69 | |
AK051639.1 | - | 9 - 10 | 2267 | 69 | ||
AK054492.1 | - | 9 - 10 | 2254 | 69 | ||
AK157802.1 | - | 8 - 9 | 3861 | 69 | ||
BC158103.1 | - | 9 - 10 | 2268 | 9088180 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
