Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700009937.mm.1 |
NM_029835.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009937.mm.1 |
NM_029835.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009937.mm.1 |
TC0700001131.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700009936.mm.1 |
NM_029835.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009936.mm.1 |
NM_029835.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009936.mm.1 |
TC0700001131.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700009938.mm.1 |
NM_029835.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009938.mm.1 |
NM_029835.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009938.mm.1 |
TC0700001131.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700009940.mm.1 |
NM_029835.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009940.mm.1 |
NM_029835.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009940.mm.1 |
TC0700001131.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700009943.mm.1 |
NM_029835.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009943.mm.1 |
NM_029835.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009943.mm.1 |
TC0700001131.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700009942.mm.1 |
NM_029835.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009942.mm.1 |
NM_029835.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009942.mm.1 |
TC0700001131.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700009944.mm.1 |
NM_029835.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009944.mm.1 |
NM_029835.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009944.mm.1 |
TC0700001131.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700009941.mm.1 |
NM_029835.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009941.mm.1 |
NM_029835.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009941.mm.1 |
TC0700001131.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700009939.mm.1 |
NM_029835.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009939.mm.1 |
NM_029835.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009939.mm.1 |
TC0700001131.mm.1 |
MTA TC |