This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01318221_m1
See other DPYD GE Assays ›
Gene Symbol:
Dpyd
Gene Name:
dihydropyrimidine dehydrogenase
Gene Aliases:
AI315208, DPD, E330028L06Rik
Chromosome Location:
Chr.3: 118562109 - 119432918 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01318221_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dpyd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_170778.2 NP_740748.1 21 - 22 2859 77
GenBank mRNA AK054474.1 - 2 - 3 285 77
BC028831.1 - 3 - 4 403 77
BC039699.1 - 21 - 22 2860 4235971 77
BC042543.1 - 17 - 18 2324 77
BC044730.2 - 9 - 10 1222 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300001221.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dpyd

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

purine nucleobase catabolic process
pyrimidine nucleobase catabolic process
thymine catabolic process
uracil catabolic process
thymidine catabolic process
UMP biosynthetic process
response to nutrient
circadian rhythm
uracil metabolic process
response to drug
oxidation-reduction process
nucleotide binding
uracil binding
catalytic activity
dihydroorotate dehydrogenase activity
oxidoreductase activity
oxidoreductase activity, acting on the CH-CH group of donors
dihydropyrimidine dehydrogenase (NADP+) activity
protein homodimerization activity
metal ion binding
NADP binding
iron-sulfur cluster binding
4 iron, 4 sulfur cluster binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300009928.mm.1 NM_170778.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300009928.mm.1 NM_170778.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300009928.mm.1 TC0300001221.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0