This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01317822_m1
See other MFGE8 GE Assays ›
Gene Symbol:
Mfge8
Gene Name:
milk fat globule-EGF factor 8 protein
Gene Aliases:
AA408458, AI325141, MFG-E8, MP47, Mfgm, P47, SED1, lactadherin
Chromosome Location:
Chr.7: 79133768 - 79149060 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01317822_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mfge8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001045489.1 NP_001038954.1 6 - 7 91
NM_008594.2 NP_032620.2 7 - 8 91
GenBank mRNA AB021130.2 - 7 - 8 91
AB025280.1 - 6 - 7 91
AK087555.1 - 6 - 7 91
AK089211.1 - 6 - 7 91
AK145183.1 - 6 - 7 91
AK148371.1 - 6 - 7 91
AK152088.1 - 7 - 8 91
AK155047.1 - 7 - 8 91
AK169997.1 - 6 - 7 91
AK170935.1 - 6 - 7 91
AK171143.1 - 7 - 8 91
BC003892.1 - 6 - 7 3593310 91
BC003904.1 - 6 - 7 3601565 91
BC018577.1 - 6 - 7 91
BC037475.1 - 3 - 4 91
M38337.1 - 7 - 8 959 91
Y11684.2 - 6 - 7 91

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mfge8

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
phagocytosis, recognition
phagocytosis, engulfment
cell adhesion
single fertilization
positive regulation of phagocytosis
positive regulation of apoptotic cell clearance
phosphatidylserine binding
integrin binding
phosphatidylethanolamine binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0