This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01317804_m1
See other AP3S2 GE Assays ›
Gene Symbol:
Ap3s2
Gene Name:
adaptor-related protein complex 3, sigma 2 subunit
Gene Aliases:
[s]3B
Chromosome Location:
Chr.7: 79875325 - 79920640 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01317804_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ap3s2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009682.3 NP_033812.3 4 - 5 409 144
GenBank mRNA AK028372.1 - 4 - 5 392 144
AK037336.1 - 4 - 5 387 144
AK083911.1 - 4 - 5 398 144
AK150085.1 - 4 - 5 402 144
AK214389.1 - 2 - 3 151 144
BC060236.1 - 4 - 5 370 6853219 144
U91933.1 - 4 - 5 364 144

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700003572.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ap3s2

Panther Classification:

Molecular Function -

vesicle coat protein

Gene Ontology Categories:

Function(s) Process(es)

transport
intracellular protein transport
anterograde axonal transport
protein transport
vesicle-mediated transport
anterograde synaptic vesicle transport
protein transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700030199.mm.1 NM_009682.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700030199.mm.1 NM_009682.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700030199.mm.1 TC0700003572.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0