This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01317684_m1
See other PPP3CA GE Assays ›
Gene Symbol:
Ppp3ca
Gene Name:
protein phosphatase 3, catalytic subunit, alpha isoform
Gene Aliases:
2900074D19Rik, CN, Caln, Calna, CnA
Chromosome Location:
Chr.3: 136670066 - 136937727 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01317684_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp3ca
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001293622.1 NP_001280551.1 8 - 9 1739 125
NM_008913.5 NP_032939.1 8 - 9 1739 125
GenBank mRNA AK146387.1 - 8 - 9 1683 125
AK150393.1 - 8 - 9 1098 125
AK166287.1 - 8 - 9 1516 126
BC138612.1 - 8 - 9 1012 8861632 125
J04134.1 - 3 - 4 319 125
J05479.1 - 8 - 9 1036 125

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300001382.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp3ca

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
protein dephosphorylation
protein import into nucleus
calcium ion transport
transition between fast and slow fiber
cardiac muscle hypertrophy in response to stress
dephosphorylation
calcium-mediated signaling
calcineurin-NFAT signaling cascade
multicellular organismal response to stress
negative regulation of chromatin binding
cellular response to drug
positive regulation of transcription from RNA polymerase II promoter
negative regulation of insulin secretion
skeletal muscle fiber development
negative regulation of dendrite morphogenesis
modulation of synaptic transmission
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of NFAT protein import into nucleus
response to calcium ion
excitatory postsynaptic potential
cellular response to glucose stimulus
positive regulation of cardiac muscle hypertrophy in response to stress
phosphoprotein phosphatase activity
protein serine/threonine phosphatase activity
calcium-dependent protein serine/threonine phosphatase activity
protein binding
calmodulin binding
drug binding
hydrolase activity
enzyme binding
calmodulin-dependent protein phosphatase activity
metal ion binding
protein heterodimerization activity
protein dimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300011189.mm.1 NM_008913.5 Refseq
Affymetrix Mouse Clariom S Array PSR0300011189.mm.1 TC0300001382.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0