This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01317464_m1
See other NUGGC GE Assays ›
Gene Symbol:
Nuggc
Gene Name:
nuclear GTPase, germinal center associated
Gene Aliases:
Gm600, SLIP-GC
Chromosome Location:
Chr.14: 65593499 - 65648444 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01317464_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nuggc
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001195674.2 NP_001182603.2 6 - 7 761 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400000937.mm.1

Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

cellular response to DNA damage stimulus
somatic hypermutation of immunoglobulin genes
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400007490.mm.1 NM_001195674.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400007490.mm.1 TC1400000937.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0