This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01317173_m1
See other DIAP3 GE Assays ›
Gene Symbol:
Diap3
Gene Name:
diaphanous homolog 3 (Drosophila)
Gene Aliases:
4930417P13Rik, Dia2, Diaph3, Drf3, mDia2, mKIAA4117, p134MDia2
Chromosome Location:
Chr.14: 86655361 - 87141248 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01317173_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Diap3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019670.1 NP_062644.1 5 - 6 565 152
GenBank mRNA AF094519.1 - 5 - 6 565 152
AK137073.1 - 4 - 5 654 152
AK138694.1 - 5 - 6 595 152
AK161737.1 - 4 - 5 583 152
AK166882.1 - 5 - 6 621 152
AK220493.1 - 5 - 6 629 152
BC003250.1 - 4 - 5 599 152
BC085191.1 - 5 - 6 675 30693873 152
BC086779.1 - 5 - 6 640 152

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400002515.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Diap3

Gene Ontology Categories:

Function(s) Process(es)

cellular component organization
actin cytoskeleton organization
actin binding
Rho GTPase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400019304.mm.1 NM_019670.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400019304.mm.1 NM_019670.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400019304.mm.1 TC1400002515.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0