This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01317047_m1
See other PPP3CC GE Assays ›
Gene Symbol:
Ppp3cc
Gene Name:
protein phosphatase 3, catalytic subunit, gamma isoform
Gene Aliases:
Calnc, PP2BA gamma
Chromosome Location:
Chr.14: 70217864 - 70289508 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01317047_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp3cc
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK015002.1 - 10 - 11 1357 90
AK133393.1 - 10 - 11 1378 90
BC141079.1 - 10 - 11 1198 9055691 90
BC145347.1 - 9 - 10 1073 90
M81475.1 - 10 - 11 1351 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400002335.mm.1

Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

protein phosphatase

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400018179.mm.1 NM_008915.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400018179.mm.1 TC1400002335.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0