Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400004167.mm.1 |
NM_001145953.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004167.mm.1 |
NM_001145953.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004167.mm.1 |
NM_010705.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004167.mm.1 |
NM_010705.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004167.mm.1 |
TC1400000513.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400004158.mm.1 |
NM_001145953.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004158.mm.1 |
NM_001145953.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004158.mm.1 |
NM_010705.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004158.mm.1 |
NM_010705.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004158.mm.1 |
TC1400000513.mm.1 |
MTA TC |