This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01316532_g1
See other TMBIM7 GE Assays ›
Gene Symbol:
Tmbim7
Gene Name:
transmembrane BAX inhibitor motif containing 7
Gene Aliases:
4930403J02Rik, 4930500J03Rik, 4930511M11Rik, Lgf5, Tmbim1b
Chromosome Location:
Chr.5: 3657004 - 3680677 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01316532_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmbim7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029141.4 NP_083417.1 7 - 8 923 82
NM_029154.1 NP_083430.1 7 - 8 689 82
GenBank mRNA AK015666.1 - 7 - 8 923 82
AK015761.1 - 7 - 8 514 82
AK161344.1 - 5 - 6 384 82
BC115681.2 - 7 - 8 781 40104180 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000017.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmbim7

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500000149.mm.1 NM_029141.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500000149.mm.1 NM_029141.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500000149.mm.1 NM_029154.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500000149.mm.1 NM_029154.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500000149.mm.1 TC0500000017.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0