This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01316087_m1
See other NFKB1 GE Assays ›
Gene Symbol:
Nfkb1
Gene Name:
nuclear factor of kappa light polypeptide gene enhancer in B cells 1, p105
Gene Aliases:
NF-KB1, NF-kappaB, NF-kappaB1, p105, p50, p50/p105
Chromosome Location:
Chr.3: 135584655 - 135691547 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01316087_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nfkb1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008689.2 NP_032715.2 17 - 18 2235 106
GenBank mRNA AB119195.1 - 15 - 16 1733 106
AB221614.1 - 15 - 16 1740 106
AK089660.1 - 5 - 6 609 106
AK146389.1 - 3 - 4 455 106
AK157915.1 - 2 - 3 235 106
AK159423.1 - 5 - 6 666 106
AY521462.1 - 15 - 16 1733 106
AY521463.1 - 15 - 16 1733 106
BC138535.1 - 16 - 17 1812 8861555 106
BC138536.1 - 16 - 17 1812 8861556 106
BC145317.1 - 17 - 18 1829 106
M57999.1 - 17 - 18 2024 106
S66656.1 - 5 - 6 779 106
S89033.1 - 5 - 6 827 106

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002994.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nfkb1

Panther Classification:

Molecular Function -

Rel homology transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
negative regulation of cytokine production
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
apoptotic process
inflammatory response
response to oxidative stress
signal transduction
I-kappaB kinase/NF-kappaB signaling
negative regulation of calcidiol 1-monooxygenase activity
response to cytokine
response to muscle stretch
NIK/NF-kappaB signaling
negative regulation of apoptotic process
negative regulation of interleukin-12 biosynthetic process
innate immune response
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
lymph node development
negative regulation of inflammatory response
cellular response to lipopolysaccharide
cellular response to nicotine
cellular response to interleukin-6
cellular response to dsRNA
cellular response to peptide hormone stimulus
positive regulation of canonical Wnt signaling pathway
positive regulation of hyaluronan biosynthetic process
positive regulation of miRNA metabolic process
regulatory region DNA binding
transcription regulatory region sequence-specific DNA binding
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
chromatin binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
heat shock protein binding
identical protein binding
protein homodimerization activity
sequence-specific DNA binding
transcription regulatory region DNA binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300023621.mm.1 NM_008689.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300023621.mm.1 NM_008689.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300023621.mm.1 TC0300002994.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0