This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01315843_m1
See other 4930592C13RIK GE Assays ›
Gene Symbol:
4930592C13Rik
Gene Name:
RIKEN cDNA 4930592C13 gene
Gene Aliases:
Gm1657
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01315843_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930592C13Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK076987.1 - 1 - 2 88 74
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB616011.1 - 1 - 2 85 Chr. 3 - 157527532 - 157534088 [-] on Build GRCm38 74

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream neuronal growth regulator 1 Negr1 5330422G01Rik,
KILON,
Ntra
-211.1kb Anti-sense Chr. 3 - 156561794 - 157316464 on NCBI Build GRCm38
Downstream zinc finger,
RAN-binding domain containing 2
Zranb2 AI227013,
Zfp265,
Zis,
Znf265
307.0 Anti-sense Chr. 3 - 157534397 - 157548339 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1442611_PM_at - -
Mouse Genome 430 2.0 Array 1442611_at - -
Mouse Genome 430 2.0 Array 1442611_PM_at - -
Mouse Expression Set 430 1442611_at - -
Mouse Expression Set 430 1442611_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1442611_at - -
Affymetrix HT Mouse Genome 430B Array 1442611_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 879980 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0