This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01315362_m1
See other SLC28A1 GE Assays ›
Gene Symbol:
Slc28a1
Gene Name:
solute carrier family 28 (sodium-coupled nucleoside transporter), member 1
Gene Aliases:
Cnt1
Chromosome Location:
Chr.7: 81113269 - 81170416 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01315362_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc28a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001004184.3 NP_001004184.2 4 - 5 368 70
GenBank mRNA BC061230.1 - 4 - 5 368 30379959 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700001168.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc28a1

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

purine nucleobase transport
pyrimidine nucleobase transport
sodium ion transmembrane transport
pyrimidine-containing compound transmembrane transport
nucleoside transmembrane transport
pyrimidine nucleobase transmembrane transporter activity
pyrimidine- and adenine-specific:sodium symporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700010260.mm.1 NM_001004184.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700010260.mm.1 NM_001004184.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700010260.mm.1 TC0700001168.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0