Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400018388.mm.1 |
NM_181345.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018388.mm.1 |
NM_181345.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018388.mm.1 |
TC1400002348.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400018387.mm.1 |
NM_181345.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018387.mm.1 |
NM_181345.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018387.mm.1 |
TC1400002348.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400018391.mm.1 |
NM_181345.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018391.mm.1 |
NM_181345.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018391.mm.1 |
TC1400002348.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400018390.mm.1 |
NM_181345.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018390.mm.1 |
NM_181345.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018390.mm.1 |
TC1400002348.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400018389.mm.1 |
NM_181345.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018389.mm.1 |
NM_181345.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018389.mm.1 |
TC1400002348.mm.1 |
MTA TC |