This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01313238_g1
See other SEC24B GE Assays ›
Gene Symbol:
Sec24b
Gene Name:
Sec24 related gene family, member B (S. cerevisiae)
Gene Aliases:
AI605202, SEC24
Chromosome Location:
Chr.3: 129982764 - 130061573 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01313238_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sec24b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_207209.2 NP_997092.1 9 - 10 2096 97
GenBank mRNA AK150076.1 - 8 - 9 1173 97
AK152339.1 - 7 - 8 1068 97
BC046556.1 - 9 - 10 1928 97
BC046776.1 - 9 - 10 2096 5706007 97
BC057355.1 - 9 - 10 1957 6833698 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002939.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sec24b

Panther Classification:

Molecular Function -

vesicle coat protein

Gene Ontology Categories:

Function(s) Process(es)

neural tube closure
auditory receptor cell morphogenesis
outflow tract morphogenesis
cochlear nucleus development
aorta morphogenesis
auditory receptor cell stereocilium organization
lung morphogenesis
lung lobe morphogenesis
coronary artery morphogenesis
pulmonary artery morphogenesis
cardiovascular system development
regulation of establishment of planar polarity involved in neural tube closure
regulation of cargo loading into COPII-coated vesicle
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300023288.mm.1 NM_207209.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300023288.mm.1 NM_207209.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300023288.mm.1 TC0300002939.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0