Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300024453.mm.1 |
NM_001164198.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024453.mm.1 |
NM_001164198.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024453.mm.1 |
NM_001164199.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024453.mm.1 |
NM_001164199.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024453.mm.1 |
NM_001164200.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024453.mm.1 |
NM_001164200.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024453.mm.1 |
NM_011100.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024453.mm.1 |
NM_011100.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024453.mm.1 |
TC0300003106.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300024460.mm.1 |
NM_011100.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024460.mm.1 |
NM_011100.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024460.mm.1 |
TC0300003106.mm.1 |
MTA TC |